Assay Details
Target Gene Details
Entrez Gene ID: | 24071 |
Gene Name: | synaptojanin 2 binding protein |
Gene Aliases: |
AA409442, ARIP2, ARIP2a, ActRIP4, D12Wsu118e, OMP25 |
Location: |
Chr.12:81492194-81532907 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Synj2bp | NM_025292.6 | NP_079568.1 | ||
AK008054.1 | BAB25432.1 | |||
AK008254.1 | 3 | 392 | ||
AK013474.1 | BAB28873.1 | |||
AK030379.1 | ||||
AK031475.1 | 1 | 2076 | ||
AK033514.1 | BAC28333.1 | |||
AK078422.1 | ||||
AK163362.1 | BAE37317.1 | |||
AY071903.1 | 2 | 695 | AAL60065.1 | |
AY138960.1 | AAN17786.1 | |||
AY566156.1 | 3 | 395 | AAT70239.1 | |
BC027433.1 | AAH27433.2 |
More Information
Set Membership: |
Intragenic Non-exonic |