Assay Details
Target Gene Details
Entrez Gene ID: | 382562 |
Gene Name: | profilin family, member 4 |
Gene Aliases: |
2900024P18Rik |
Location: |
Chr.12:4768913-4778808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pfn4 | NM_028376.3 | NP_082652.1 | ||
AK013595.1 | BAB28919.1 | |||
AY495949.1 | AAS75143.1 | |||
BC116888.1 | ||||
BC116890.1 |
More Information
Set Membership: |
Intragenic Non-exonic |