Assay Details
Target Gene Details
Entrez Gene ID: | 17252 |
Gene Name: | retinol dehydrogenase 11 |
Gene Aliases: |
2610319N22Rik, AI428145, AU045252, Arsdr1, C85936, CGI-82, HCBP12, M42C60, Mdt1, Psdr1, SCALD, UBE-1c1, Ube-1c |
Location: |
Chr.12:79174335-79191819 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rdh11 | NM_021557.5 | 5 | 534 | NP_067532.2 |
AB030503.1 | 5 | 866 | BAA82656.1 | |
AB030504.1 | 5 | 614 | BAA82657.1 | |
AB035959.1 | 5 | 542 | BAA88521.1 | |
AF474027.1 | 5 | 580 | AAL79910.1 | |
AK004413.1 | 5 | 535 | BAB23296.1 | |
AK012056.1 | 5 | 562 | ||
AK135443.1 | 5 | 564 | BAE22534.1 | |
AY039032.1 | 5 | 1008 | AAK91516.1 | |
BC018261.1 | 5 | 559 | AAH18261.1 |
More Information
Set Membership: |
Intragenic Exonic |