Assay Details
Target Gene Details
Entrez Gene ID: | 11992 |
Gene Name: | AU RNA binding protein/enoyl-coenzyme A hydratase |
Gene Aliases: |
C77140, W91705 |
Location: |
Chr.13:52835102-52929706 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Auh | NM_016709.2 | NP_057918.2 | ||
AK003219.1 | ||||
AK003929.1 | BAB23078.1 | |||
AK019978.1 | BAB31947.1 | |||
AK134886.1 | ||||
AK147855.1 | ||||
AK169661.1 | BAE41283.1 | |||
BC026525.1 | AAH26525.1 | |||
BC049597.1 | AAH49597.1 | |||
BY741014.1 |
More Information
Set Membership: |
Intragenic Non-exonic |