Assay Details
Target Gene Details
Entrez Gene ID: | 12192 |
Gene Name: | zinc finger protein 36, C3H type-like 1 |
Gene Aliases: |
AW742437, AW743212, Berg36, Brf1, D530020L18Rik, ERF1, TIS11b, cMG1 |
Location: |
Chr.12:80107760-80113013 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zfp36l1 | NM_007564.5 | NP_031590.1 | ||
AK021295.1 | 1 | 184 | ||
AK036950.1 | BAC29648.1 | |||
AK051035.1 | BAC34506.1 | |||
AK077855.1 | ||||
BC016621.1 | AAH16621.1 | |||
M58566.1 | AAA72948.1 |
More Information
Set Membership: |
Intragenic Non-exonic |