Assay Details
Target Gene Details
Entrez Gene ID: | 83602 |
Gene Name: | general transcription factor II A, 1 |
Gene Aliases: |
6330549H03Rik, AA536742, AA959775, AW060250, TfIIAa/b, Tfiia1 |
Location: |
Chr.12:91555262-91590487 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gtf2a1 | NM_031391.2 | NP_113568.2 | ||
NM_175335.3 | NP_780544.1 | |||
AF250834.1 | AAG50431.1 | |||
AK048711.1 | BAC33430.1 | |||
BC079909.1 | ||||
BC117829.1 | ||||
BC117830.1 |
More Information
Set Membership: |
Intragenic Non-exonic |