Assay Details
Target Gene Details
Entrez Gene ID: | 12454 |
Gene Name: | cyclin K |
Gene Aliases: |
AW123198, AW413594, CPR4, CycK |
Location: |
Chr.12:108179446-108203362 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnk | NM_009832.2 | 3 | 339 | NP_033962.2 |
AF060517.1 | 1 | 108 | AAD09979.1 | |
AK089386.1 | 3 | 276 | ||
AK146711.1 | 3 | 276 | BAE27377.1 | |
AK154679.1 | 3 | 276 | BAE32760.1 | |
AK154765.1 | 3 | 276 | BAE32814.1 | |
BC027297.1 | 3 | 234 | AAH27297.1 | |
CA984404.1 | 2 | 465 |
More Information
Set Membership: |
Intragenic Exonic |