Assay Details
Target Gene Details
Entrez Gene ID: | 12454 |
Gene Name: | cyclin K |
Gene Aliases: |
AW123198, AW413594, CPR4, CycK |
Location: |
Chr.12:108179446-108203362 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccnk | NM_009832.2 | 1 | 67 | NP_033962.2 |
AK089386.1 | ||||
AK146711.1 | BAE27377.1 | |||
AK154679.1 | BAE32760.1 | |||
AK154765.1 | BAE32814.1 | |||
BC027297.1 | AAH27297.1 | |||
CA984404.1 | 1 | 225 |
More Information
Set Membership: |
Intragenic Exonic |