Assay Details
Target Gene Details
Entrez Gene ID: | 18109 |
Gene Name: | v-myc myelocytomatosis viral related oncogene, neuroblastoma derived (avian) |
Gene Aliases: |
N-myc, Nmyc, Nmyc-1, Nmyc1, bHLHe37, c-nmyc |
Location: |
Chr.12:12936093-12941836 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mycn | NM_008709.3 | NP_032735.3 | ||
AK144126.1 | ||||
AK146696.1 | BAE27366.1 | |||
AK146913.1 | BAE27525.1 |
More Information
Set Membership: |
Intragenic Non-exonic |