Assay Details
Target Gene Details
Entrez Gene ID: | 208439 |
Gene Name: | kelch-like 29 |
Gene Aliases: |
A230106N14Rik, Gm68, Kbtbd9, mKIAA1921 |
Location: |
Chr.12:5077465-5375682 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhl29 | NM_001164493.1 | NP_001157965.1 | ||
AK122572.1 | BAC65854.1 | |||
BC138283.1 | ||||
BC145230.1 | ||||
BC145748.1 |
More Information
Set Membership: |
Intragenic Non-exonic |