Assay Details
Target Gene Details
Entrez Gene ID: | 18263 |
Gene Name: | ornithine decarboxylase, structural 1 |
Gene Aliases: |
ODC |
Location: |
Chr.12:17544873-17551502 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Odc1 | NM_013614.2 | NP_038642.2 | ||
AK077679.1 | ||||
AK139610.1 | BAE24082.1 | |||
AK150283.1 | ||||
AK150343.1 | ||||
AK152525.1 | ||||
AK166093.1 | BAE38566.1 | |||
AK168219.1 | ||||
AK169214.1 | ||||
AK172568.1 | ||||
AK190288.1 | ||||
AK194223.1 | ||||
AK194907.1 | ||||
AK201555.1 | ||||
AK202843.1 | ||||
BC033264.1 | AAH33264.1 | |||
BC059826.1 | AAH59826.1 | |||
BC083122.1 | AAH83122.1 | |||
EU684749.1 | ||||
M12330.1 | AAA39846.1 | |||
M12331.1 | AAA39848.1 | |||
M20617.2 | AAA51638.1 | |||
S64539.1 | AAB27809.1 |
More Information
Set Membership: |
Intragenic Non-exonic |