Assay Details
Target Gene Details
Entrez Gene ID: | 66174 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 14 |
Gene Aliases: |
1110030M18Rik |
Location: |
Chr.12:112934727-112942167 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nudt14 | NM_025399.3 | NP_079675.1 | ||
AK003991.1 | BAB23110.1 | |||
AK013174.1 | BAB28691.2 | |||
BC025444.1 | AAH25444.1 |
More Information
Set Membership: |
Intragenic Non-exonic |