Assay Details
Target Gene Details
Entrez Gene ID: | 17977 |
Gene Name: | nuclear receptor coactivator 1 |
Gene Aliases: |
KAT13A, SRC-1, SRC1, bHLHe74 |
Location: |
Chr.12:4247362-4477210 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncoa1 | NM_010881.2 | NP_035011.1 | ||
BC068177.1 | AAH68177.1 | |||
BC080866.1 | AAH80866.1 | |||
U56920.1 | AAB01228.1 | |||
U64606.1 | AAB06177.1 | |||
U64828.1 | AAB38841.1 |
More Information
Set Membership: |
Intragenic Non-exonic |