Assay Details
Target Gene Details
Entrez Gene ID: | 268566 |
Gene Name: | gephyrin |
Gene Aliases: |
5730552E08Rik, AI662856, BC027112, C230040D23, GPH, GPHRYN, geph |
Location: |
Chr.12:78224153-78684772 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gphn | NM_145965.2 | NP_666077.2 | ||
NM_172952.3 | NP_766540.2 | |||
AK082353.1 | BAC38476.1 | |||
AK144569.1 | ||||
AK183551.1 | ||||
BC127058.1 |