Assay Details
Target Gene Details
Entrez Gene ID: | 72308 |
Gene Name: | BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB (S. cerevisiae) |
Gene Aliases: |
2510002F24Rik, GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90 |
Location: |
Chr.12:112955631-113000707 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Brf1 | NM_028193.3 | NP_082469.2 | ||
BC037224.1 | AAH37224.1 | |||
BU148121.1 | ||||
CF737042.1 |
Target Gene Details
Entrez Gene ID: | 399566 |
Gene Name: | BTB (POZ) domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.12:112974998-112979493 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Btbd6 | NM_001145900.1 | NP_001139372.1 | ||
NM_201646.2 | NP_964008.2 | |||
AI119004.1 | ||||
AK146991.1 | BAE27593.1 | |||
BC031195.1 | AAH31195.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |