Assay Details
Target Gene Details
Entrez Gene ID: | 320817 |
Gene Name: | ATPase family, AAA domain containing 2B |
Gene Aliases: |
1110014E10Rik, BC032887, C79189, D530031C13Rik |
Location: |
Chr.12:4916084-5047410 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atad2b | NM_001099628.1 | NP_001093098.1 | ||
AK080580.1 | ||||
AK085232.1 | BAC39396.1 | |||
AK161170.1 | 4 | 885 |
More Information
Set Membership: |
Intragenic Non-exonic |