Assay Details
Target Gene Details
Entrez Gene ID: | 23876 |
Gene Name: | fibulin 5 |
Gene Aliases: |
A55, DANCE, EVEC |
Location: |
Chr.12:101746565-101819130 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln5 | NM_011812.4 | 8 | 1173 | NP_035942.1 |
AF112151.1 | 8 | 1123 | AAD41767.1 | |
AK085170.1 | 8 | 1003 | BAC39381.1 | |
AK090129.1 | 8 | 1112 | BAC41109.1 | |
AK159189.1 | 8 | 937 | ||
AK159471.1 | 8 | 1156 | BAE35112.1 | |
AK159960.1 | 8 | 1143 | BAE35515.1 | |
AK165018.1 | 8 | 966 | BAE38002.1 | |
AK180395.1 | ||||
BC006636.1 | 8 | 863 | AAH06636.1 |
More Information
Set Membership: |
Intragenic Exonic |