Assay Details
Target Gene Details
Entrez Gene ID: | 217874 |
Gene Name: | retinal degeneration 3-like |
Gene Aliases: |
A930011N13 |
Location: |
Chr.12:111979191-111981128 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rd3l | NM_001127685.1 | NP_001121157.1 | ||
AK080710.1 | 1 | 335 | ||
BC147034.1 | ||||
BC147035.1 | ||||
BC147747.1 | ||||
BC147749.1 |
Target Gene Details
Entrez Gene ID: | 74691 |
Gene Name: | tudor domain containing 9 |
Gene Aliases: |
4930441E05Rik |
Location: |
Chr.12:111971474-112068854 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tdrd9 | NM_029056.1 | NP_083332.1 | ||
AB362563.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |