Assay Details
Target Gene Details
Entrez Gene ID: | 75302 |
Gene Name: | additional sex combs like 2 (Drosophila) |
Gene Aliases: |
4930556B16Rik, mKIAA1685 |
Location: |
Chr.12:3426857-3506852 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Asxl2 | NM_001270988.1 | 8 | 1166 | NP_001257917.1 |
NM_172421.5 | 7 | 1022 | NP_766009.2 | |
AK016141.1 | 7 | 728 | ||
AK035679.1 | 2 | 1594 | ||
AK036839.1 | 7 | 979 | BAC29602.1 | |
AK080679.1 | 8 | 1140 | BAC37978.2 | |
AK148015.1 | 7 | 1027 | ||
BC141140.1 | 8 | 1000 |