Assay Details
Target Gene Details
Entrez Gene ID: | 17187 |
Gene Name: | Max protein |
Gene Aliases: |
AA960152, AI875693, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8 |
Location: |
Chr.12:76937269-76962335 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Max | NM_001146176.1 | NP_001139648.1 | ||
NM_008558.2 | NP_032584.2 | |||
AK080431.1 | BAC37914.1 | |||
AK178097.1 | ||||
AK189653.1 | ||||
BC138671.1 | ||||
BC138672.1 | ||||
BC145369.1 | ||||
M63903.1 | AAA39797.1 | |||
S80521.1 | AAB35748.1 |
More Information
Set Membership: |
Intragenic Non-exonic |