Assay Details
Target Gene Details
Entrez Gene ID: | 23876 |
Gene Name: | fibulin 5 |
Gene Aliases: |
A55, DANCE, EVEC |
Location: |
Chr.12:101746565-101819130 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fbln5 | NM_011812.4 | NP_035942.1 | ||
AF112151.1 | AAD41767.1 | |||
AK084895.1 | ||||
AK085170.1 | BAC39381.1 | |||
AK090129.1 | BAC41109.1 | |||
AK159189.1 | ||||
AK159471.1 | BAE35112.1 | |||
AK159960.1 | BAE35515.1 | |||
AK165018.1 | BAE38002.1 | |||
AK178699.1 | ||||
AK208765.1 | ||||
AK219368.1 | ||||
BC006636.1 | AAH06636.1 | |||
BQ557120.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |