Assay Details
Target Gene Details
Entrez Gene ID: | 27386 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
4930423H22Rik, bHLHe12 |
Location: |
Chr.12:53248003-54072175 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Npas3 | NM_013780.2 | NP_038808.2 | ||
AF168769.1 | AAG35181.1 | |||
AF173871.1 | AAF14283.1 | |||
BC120885.1 |
More Information
Set Membership: |
Intragenic Non-exonic |