Assay Details
Target Gene Details
Entrez Gene ID: | 52635 |
Gene Name: | extended synaptotagmin-like protein 2 |
Gene Aliases: |
2410017M09Rik, 4921504I16Rik, CHR2SYT, D12Ertd551e, Fam62b |
Location: |
Chr.12:116279680-116390935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Esyt2 | NM_028731.5 | NP_083007.2 | ||
AK014813.1 | BAB29565.1 | |||
AK044360.1 | BAC31884.1 | |||
AK157352.1 | BAE34059.1 | |||
BC052440.1 | AAH52440.1 | |||
BC059230.1 | AAH59230.1 | |||
BC138937.1 |