Assay Details
Target Gene Details
Entrez Gene ID: | 67217 |
Gene Name: | L-3-hydroxyproline dehydratase (trans-) |
Gene Aliases: |
2810055F11Rik |
Location: |
Chr.12:72071660-72085433 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
L3hypdh | NM_026038.2 | 3 | 898 | NP_080314.1 |
AK012954.1 | 3 | 1024 | ||
AK018449.1 | 3 | 899 | BAB31217.1 | |
AK137154.1 | 3 | 986 | ||
BC004753.1 | 3 | 958 | AAH04753.1 |
More Information
Set Membership: |
Intragenic Exonic |