Assay Details
Target Gene Details
Entrez Gene ID: | 238055 |
Gene Name: | apolipoprotein B |
Gene Aliases: |
AI315052, Apo B-100, apob-100, apob-48 |
Location: |
Chr.12:7977633-8016839 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Apob | NM_009693.2 | 9 | 1156 | NP_033823.2 |
BC038263.1 | 9 | 1171 | AAH38263.1 |
More Information
Set Membership: |
Intragenic Exonic |