Assay Details
Target Gene Details
Entrez Gene ID: | 20848 |
Gene Name: | signal transducer and activator of transcription 3 |
Gene Aliases: |
1110034C02Rik, AW109958, Aprf |
Location: |
Chr.11:100886810-100939570 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stat3 | NM_011486.4 | NP_035616.1 | ||
NM_213659.2 | NP_998824.1 | |||
NM_213660.2 | NP_998825.1 | |||
AK145486.1 | BAE26464.1 | |||
AK153005.1 | BAE31645.1 | |||
AK153472.1 | BAE32023.1 | |||
AY299489.1 | AAQ75418.1 | |||
AY299490.1 | AAQ75419.1 | |||
BC003806.1 | AAH03806.1 | |||
BC019168.1 | AAH19168.1 | |||
L29278.1 | AAA37254.1 | |||
U06922.1 | AAA19452.1 | |||
U08378.1 | AAA56668.1 | |||
U30709.1 | AAC52612.1 |
More Information
Set Membership: |
Intragenic Non-exonic |