Assay Details
Target Gene Details
Entrez Gene ID: | 20848 |
Gene Name: | signal transducer and activator of transcription 3 |
Gene Aliases: |
1110034C02Rik, AW109958, Aprf |
Location: |
Chr.11:100886810-100939570 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stat3 | NM_011486.4 | 6 | 794 | NP_035616.1 |
NM_213659.2 | 6 | 794 | NP_998824.1 | |
NM_213660.2 | 6 | 794 | NP_998825.1 | |
AK145486.1 | 6 | 650 | BAE26464.1 | |
AK153005.1 | 6 | 706 | BAE31645.1 | |
AK153472.1 | 6 | 706 | BAE32023.1 | |
AY299489.1 | 5 | 521 | AAQ75418.1 | |
AY299490.1 | 5 | 521 | AAQ75419.1 | |
BC003806.1 | 6 | 682 | AAH03806.1 | |
BC019168.1 | 6 | 698 | AAH19168.1 | |
L29278.1 | 6 | 773 | AAA37254.1 | |
U06922.1 | 6 | 583 | AAA19452.1 | |
U08378.1 | 6 | 661 | AAA56668.1 | |
U30709.1 | 6 | 687 | AAC52612.1 |
More Information
Set Membership: |
Intragenic Exonic |