Assay Details
Target Gene Details
Entrez Gene ID: | 11782 |
Gene Name: | adaptor-related protein complex AP-4, sigma 1 |
Gene Aliases: |
AI314282 |
Location: |
Chr.12:51689888-51738939 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap4s1 | NM_021710.3 | 2 | 403 | NP_068356.1 |
AF092093.1 | 2 | 332 | AAD20447.1 | |
AK005283.1 | 2 | 153 | BAB23931.1 | |
AK178071.1 | ||||
BC001985.1 | 2 | 403 | AAH01985.1 | |
BC053339.1 | 2 | 163 | AAH53339.1 |
More Information
Set Membership: |
Intragenic Exonic |