Assay Details
Target Gene Details
Entrez Gene ID: | 67128 |
Gene Name: | ubiquitin-conjugating enzyme E2G 1 |
Gene Aliases: |
2700059C12Rik, AI256795, AU014992, AW552068, D130023C12Rik |
Location: |
Chr.11:72607261-72686481 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ube2g1 | NM_025985.4 | NP_080261.2 | ||
AK012539.1 | ||||
AK013902.1 | BAB29048.1 | |||
AK083029.1 | ||||
AK167472.1 | BAE39555.1 | |||
AK197264.1 | ||||
DV657669.1 |