Assay Details
Target Gene Details
Entrez Gene ID: | 68729 |
Gene Name: | tripartite motif-containing 37 |
Gene Aliases: |
1110032A10Rik, 2810004E07Rik, AI848587, AU043018, MUL, TEF3, mKIAA0898 |
Location: |
Chr.11:87127051-87250874 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trim37 | NM_197987.2 | 2 | 468 | NP_932104.1 |
AB093271.1 | 2 | 455 | BAC41455.1 | |
AK012668.1 | 2 | 334 | ||
AK140822.1 | 2 | 469 | BAE24489.1 | |
BC058678.1 | 2 | 466 | AAH58678.1 | |
BC059070.1 | 2 | 434 | AAH59070.1 |
More Information
Set Membership: |
Intragenic Exonic |