Assay Details
Target Gene Details
Entrez Gene ID: | 217732 |
Gene Name: | CLOCK interacting protein, circadian |
Gene Aliases: |
2310044G17Rik, AI842544, AI853744, Kiaa1737, mKIAA1737 |
Location: |
Chr.12:86947041-86965364 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cipc | NM_001289429.1 | NP_001276358.1 | ||
NM_001289430.1 | NP_001276359.1 | |||
NM_001289431.1 | NP_001276360.1 | |||
NM_001289432.1 | NP_001276361.1 | |||
NM_173735.3 | NP_776096.2 | |||
AK122546.1 | BAC65828.1 | |||
AK154476.1 | BAE32612.1 | |||
AK160481.1 | ||||
AK160643.1 | BAE35935.1 | |||
AK165964.1 | ||||
BC026384.1 | AAH26384.1 | |||
BY097396.1 | ||||
BY287209.1 | ||||
CF530959.1 | ||||
CK391711.1 | ||||
CN526809.1 |