Assay Details
Target Gene Details
Entrez Gene ID: | 574428 |
Gene Name: | zinc finger, MYND-type containing 15 |
Gene Aliases: |
- |
Location: |
Chr.11:70459622-70466207 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zmynd15 | NM_001029929.2 | 5 | 1561 | NP_001025100.1 |
AK029988.1 | 5 | 1562 | BAC26719.1 | |
AK128979.1 | 4 | 1868 | BAC87664.1 | |
AK171931.1 | 5 | 1410 | BAE42737.1 | |
BC151086.1 | 5 | 1233 |