Assay Details
Target Gene Details
Entrez Gene ID: | 108903 |
Gene Name: | tubulin-specific chaperone d |
Gene Aliases: |
2310057L06Rik, A030005L14Rik, mKIAA0988 |
Location: |
Chr.11:121450584-121617170 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tbcd | NM_029878.3 | NP_084154.1 | ||
AK041464.1 | BAC30951.1 | |||
AK050199.1 | ||||
AK054343.1 | ||||
AK194328.1 | ||||
AK196627.1 | ||||
AK196683.1 | ||||
AK219303.1 | ||||
BC059843.1 | AAH59843.1 |
More Information
Set Membership: |
Intragenic Non-exonic |