Assay Details
Target Gene Details
Entrez Gene ID: | 217154 |
Gene Name: | SH3 and cysteine rich domain 2 |
Gene Aliases: |
24b2, 24b2/STAC2, AW240854 |
Location: |
Chr.11:98036624-98053462 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stac2 | NM_146028.4 | NP_666140.1 | ||
AJ608761.1 | CAE75538.1 | |||
AK078132.1 | BAC37142.1 | |||
AK079357.1 | BAC37621.1 | |||
AK144433.1 | BAE25886.1 | |||
BC024864.1 | AAH24864.1 | |||
BC029794.1 | AAH29794.1 | |||
BC054100.1 | AAH54100.1 |