Assay Details
Target Gene Details
Entrez Gene ID: | 217012 |
Gene Name: | unc-45 homolog B (C. elegans) |
Gene Aliases: |
AA445617, Cmya4, D230041A13Rik, Unc45 |
Location: |
Chr.11:82911253-82943406 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc45b | NM_178680.4 | 2 | 196 | NP_848795.3 |
AF539793.1 | 1 | 89 | AAO13383.1 | |
AK052294.1 | 2 | 123 | BAC34921.1 | |
AK081821.1 | 2 | 135 | BAC38342.1 | |
AK084418.1 | 2 | 164 | BAC39178.1 | |
AK168968.1 | 2 | 164 | ||
AK169274.1 | 2 | 135 | ||
BC084585.1 | 2 | 129 | AAH84585.1 | |
BY133112.1 | 2 | 196 |