Assay Details
Target Gene Details
Entrez Gene ID: | 20913 |
Gene Name: | syntaxin binding protein 4 |
Gene Aliases: |
6030470M02Rik, Synip |
Location: |
Chr.11:90476492-90638123 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stxbp4 | NM_011505.2 | 6 | 596 | NP_035635.1 |
AF152924.1 | 6 | 586 | AAD43533.1 | |
AK012293.1 | 6 | 554 | ||
AK049001.1 | 6 | 565 | ||
BC032881.1 | 6 | 550 | AAH32881.1 | |
BY720472.1 |