Assay Details
Target Gene Details
Entrez Gene ID: | 237860 |
Gene Name: | slingshot homolog 2 (Drosophila) |
Gene Aliases: |
SSH-2, SSH-2L, mSSH-2L |
Location: |
Chr.11:77216169-77460220 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ssh2 | NM_001291190.1 | NP_001278119.1 | ||
NM_177710.4 | NP_808378.2 | |||
AB099288.1 | BAC97811.1 | |||
AK155232.1 | BAE33137.1 | |||
AK170142.1 | BAE41593.1 | |||
BC141392.1 | ||||
BC141393.1 |
More Information
Set Membership: |
Intragenic Non-exonic |