Assay Details
Target Gene Details
Entrez Gene ID: | 100503361 |
Gene Name: | transmembrane protein 95 |
Gene Aliases: |
432576 |
Location: |
Chr.11:69876054-69878173 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem95 |