Assay Details
Target Gene Details
Entrez Gene ID: | 68832 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
1110057K04Rik |
Location: |
Chr.12:8208100-8299566 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ldah | NM_001167767.1 | NP_001161239.1 | ||
NM_001167768.1 | 6 | 892 | NP_001161240.1 | |
NM_172401.4 | NP_765989.3 | |||
AK076460.1 | BAC36352.1 | |||
AK079099.1 | BAC37539.1 | |||
AK086717.1 | 5 | 708 | ||
AK162850.1 | BAE37083.1 | |||
AK162914.1 | 6 | 873 | ||
AW049235.1 | 1 | 315 | ||
BC046986.1 | AAH46986.1 | |||
BC057311.1 | AAH57311.1 |
More Information
Set Membership: |
Intragenic Exonic |