Assay Details
Target Gene Details
Entrez Gene ID: | 68832 |
Gene Name: | lipid droplet associated hydrolase |
Gene Aliases: |
1110057K04Rik |
Location: |
Chr.12:8208100-8299566 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ldah | NM_001167767.1 | 3 | 340 | NP_001161239.1 |
NM_001167768.1 | 3 | 329 | NP_001161240.1 | |
NM_172401.4 | 3 | 348 | NP_765989.3 | |
AI746795.1 | 3 | 340 | ||
AK036177.1 | 3 | 318 | ||
AK076460.1 | BAC36352.1 | |||
AK079099.1 | 3 | 332 | BAC37539.1 | |
AK086717.1 | 3 | 318 | ||
AK162850.1 | BAE37083.1 | |||
AK162914.1 | 3 | 310 | ||
BB626438.1 | 3 | 329 | ||
BC046986.1 | 2 | 267 | AAH46986.1 | |
BC057311.1 | 3 | 320 | AAH57311.1 | |
BY016651.1 |
More Information
Set Membership: |
Intragenic Exonic |