Assay Details
Target Gene Details
Entrez Gene ID: | 22248 |
Gene Name: | unc-119 homolog (C. elegans) |
Gene Aliases: |
D11Bhm52, HRG4, MRG4, Rg4, Rtg4, Unc119h |
Location: |
Chr.11:78343443-78349164 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Unc119 | NM_011676.2 | NP_035806.1 | ||
AF030169.1 | AAD01893.1 | |||
AK131955.1 | BAE20899.1 | |||
BC001990.1 | AAH01990.1 |
More Information
Set Membership: |
Intragenic Non-exonic |