Assay Details
Target Gene Details
Entrez Gene ID: | 211652 |
Gene Name: | WW, C2 and coiled-coil domain containing 1 |
Gene Aliases: |
AA408228, AU017197, BC037006, Kibra, mKIAA0869 |
Location: |
Chr.11:35839178-35980089 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 23 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wwc1 | NM_170779.1 | NP_740749.1 | ||
AB271907.1 | ||||
BC006733.1 | AAH06733.1 | |||
BC017638.1 | AAH17638.1 | |||
BC037006.1 | AAH37006.1 | |||
DQ256090.1 | ABB51169.1 |