Assay Details
Target Gene Details
Entrez Gene ID: | 216965 |
Gene Name: | TAO kinase 1 |
Gene Aliases: |
2810468K05Rik, AU020252, D130018F14Rik, Map3k16, Markk, Psk2, mKIAA1361 |
Location: |
Chr.11:77529162-77607860 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Taok1 | NM_144825.2 | NP_659074.2 | ||
AK143530.1 | ||||
BC151012.1 | ||||
BC151015.1 | ||||
BC151016.1 | ||||
CF164486.1 | ||||
CN721819.1 | ||||
CN724017.1 |
More Information
Set Membership: |
Intragenic Non-exonic |