Assay Details
Target Gene Details
Entrez Gene ID: | 18412 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, OSF-6, Osi, STAP, STONE14, p62 |
Location: |
Chr.11:50200152-50210820 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sqstm1 | NM_001290769.1 | NP_001277698.1 | ||
NM_011018.3 | NP_035148.1 | |||
BC006019.1 | AAH06019.1 | |||
CX240486.1 | ||||
E08146.1 | ||||
U40930.1 | AAB17127.1 | |||
U57413.1 | AAB02908.1 |