Assay Details
Target Gene Details
Entrez Gene ID: | 216965 |
Gene Name: | TAO kinase 1 |
Gene Aliases: |
2810468K05Rik, AU020252, D130018F14Rik, Map3k16, Markk, Psk2, mKIAA1361 |
Location: |
Chr.11:77529162-77607860 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Taok1 | NM_144825.2 | 16 | 2547 | NP_659074.2 |
BC016522.1 | 4 | 541 | AAH16522.1 | |
BC151012.1 | 16 | 2341 | ||
BC151015.1 | 16 | 2341 | ||
BC151016.1 | 16 | 2340 | ||
CN717576.1 | 3 | 355 |
More Information
Set Membership: |
Intragenic Exonic |