Assay Details
Target Gene Details
Entrez Gene ID: | 20851 |
Gene Name: | signal transducer and activator of transcription 5B |
Gene Aliases: |
- |
Location: |
Chr.11:100780731-100859004 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stat5b | NM_001113563.1 | 14 | 1926 | NP_001107035.1 |
NM_011489.3 | 14 | 2258 | NP_035619.3 | |
AK045531.1 | 14 | 2069 | ||
AK150098.1 | 14 | 1895 | BAE29305.1 | |
AK154014.1 | 14 | 2258 | BAE32317.1 | |
AK154664.1 | 14 | 1892 | BAE32752.1 | |
AY040231.1 | 14 | 2212 | AAK74074.1 | |
AY042906.1 | 14 | 2212 | AAL05590.1 | |
AY044901.1 | 14 | 2212 | AAK97791.1 | |
AY044902.1 | 14 | 2212 | AAK97792.1 | |
AY044903.1 | 14 | 2212 | AAK97793.1 | |
BC024319.1 | 14 | 1882 | AAH24319.1 | |
U21110.2 | 14 | 2212 | AAC52282.1 | |
Z48539.1 | 14 | 1826 | CAA88420.1 |
More Information
Set Membership: |
Intragenic Exonic |