Assay Details
Target Gene Details
Entrez Gene ID: | 66176 |
Gene Name: | N-acetyltransferase 9 (GCN5-related, putative) |
Gene Aliases: |
1110028N05Rik |
Location: |
Chr.11:115182831-115187886 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nat9 |
Target Gene Details
Entrez Gene ID: | 320534 |
Gene Name: | transmembrane protein 104 |
Gene Aliases: |
C630005D06Rik, mFLJ00021 |
Location: |
Chr.11:115187487-115247025 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tmem104 | NM_001033393.2 | NP_001028565.1 | ||
AK040894.1 | ||||
AK049868.1 | ||||
AK131112.1 | BAD21362.1 | |||
AK155041.1 | BAE33008.1 | |||
AK167098.1 | ||||
AK171314.1 | ||||
AK171457.1 | BAE42466.1 | |||
BC140976.1 | ||||
BY091433.1 |