Assay Details
Target Gene Details
Entrez Gene ID: | 327942 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class L |
Gene Aliases: |
Gm737 |
Location: |
Chr.11:62458460-62513900 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pigl | NM_001039536.2 | NP_001034625.1 | ||
AK128932.1 | BAC87658.1 | |||
AK158149.1 | BAE34385.1 | |||
BC096678.1 | 1 | 212 | AAH96678.1 |
More Information
Set Membership: |
Intragenic Non-exonic |