Assay Details
Target Gene Details
Entrez Gene ID: | 66753 |
Gene Name: | endoplasmic reticulum lectin 1 |
Gene Aliases: |
4933407N01Rik |
Location: |
Chr.11:30865632-30954173 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Erlec1 |
Target Gene Details
Entrez Gene ID: | 103554 |
Gene Name: | proteasome (prosome, macropain) activator subunit 4 |
Gene Aliases: |
AA409398, AU041366, TEMO, mKIAA0077 |
Location: |
Chr.11:30771328-30881150 on Build GRCm38 |
Assay Gene Location: | Within Exon 44 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psme4 | NM_134013.3 | 44 | 5430 | NP_598774.2 |
AF296169.1 | 44 | 5141 | AAG09060.1 | |
AK129049.1 | 3 | 3410 | BAC97859.1 | |
AK131932.1 | BAE20881.1 | |||
AK169447.1 | 3 | 327 | BAE41181.1 | |
BC004575.1 | 2 | 259 | AAH04575.1 | |
BC031174.1 | 16 | 1874 | AAH31174.1 | |
BC054364.1 | 2 | 176 | AAH54364.1 | |
BC141381.1 | 44 | 5143 |