Assay Details
Target Gene Details
Entrez Gene ID: | 237625 |
Gene Name: | phospholipase A2, group III |
Gene Aliases: |
- |
Location: |
Chr.11:3487596-3499346 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pla2g3 | NM_172791.2 | NP_766379.2 | ||
AK033572.1 | BAC28367.1 | |||
AK046215.1 | 3 | 1483 | BAC32640.1 | |
AK081075.1 | BAC38128.1 | |||
BC079556.1 | AAH79556.1 |